VWR® 10 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR International
Certificates
About this item
10-425 autosampler screw thread vials feature a wide opening compatible with 6 mm inserts.
- Choose from all-steel or improved polymeric-metal version
- Both compatible with all standard or magnetic Headspace autosamplers
- Excellent sealing characteristics that withstand high pressures of headspace applications
Spring promotes alignment and ensures insert seals against septa to help guard against evaporation loss.
Specifications
- Catalog No:
- 82028-426A
- 89523-484A
- 89523-482A
- 82028-432A
- 82028-428A
- Kit:
- 89523-484A
- 89523-482A
- Component vial:
- 82028-432A
- 82028-428A
- Component cap:
- 82028-434A
- 82028-434A
Specifications
Caps
Product Information
Frequently Bought Together