Power supply adapters
Supplier:   Cytiva
Power supply adapters
Supplier:  Cytiva
Certificates
Power supply adapters
2 Options Available Below
Product Details & Documents
Choice of adapters for connecting 4 or 2 mm leads to 2 or 4 mm output jacks, respectively.
  • Approved for 500 V relative protection ground
  • Adapter set 2 mm M/4 mm F for connecting sheathed 2 mm male plug instrument leads to 4 mm sheathed output jacks
  • Adapter set 4 mm M/2 mm F for connecting sheathed 4 mm male plug instrument leads to 2 mm high voltage output jacks
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Adapter Set 4 mm M/2 mm F
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $315.03
    $315.03
    Each
       $315.03
    Supplier Number: 18-1129-58
  • Adapter Set 2 mm M/4 mm F
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $216.66
    $216.66
    Each
       $216.66
    Supplier Number: 18-1129-59