Specimen Containers with Caps, Caplugs
: Caplugs
Free standing containers that are ideal for collecting/transporting general purpose specimens. The containers are constructed of high-clarity polypropylene copolymer with high density polyethylene screw caps.
- All containers are sold with caps
- All containers are available with patient information labels
- All containers are natural color
Designed for storing biological fluids and pathology specimens.
Frequently Bought Together