Five-Slide Mailer, Caplugs | Avantor
Five-Slide Mailer, Caplugs
Polypropylene slide mailer for 5 slides, Natural
Catalog # 76502-820
Supplier:   Caplugs
Fournisseur #:  240-3074-030
Five-Slide Mailer, Caplugs
Catalog # 76502-820
Supplier:  Caplugs
Five-Slide Mailer, Caplugs
Five-Slide Mailer, Caplugs
Polypropylene slide mailer for 5 slides, Natural
Price
51,68 $
Log in pour la disponibilité
Carton of 100
Product Details & Documents
The Five-Slide Mailer features an integral recessed lid design which limits slide movement during transit to prevent accidental breakage.
  • Rounded corners allow easier handling
  • Eyelet is provided for optional security ties and/or identification tags
  • Mailers can also function as miniature Coplin jars
Lightweight design (less than ¹/₃ oz.) helps to minimize transport costs.
Spécifications
  • Color:
    Natural
  • Désignation:
    Polypropylene slide mailer for 5 slides