Ward's® Breccia | Avantor
Ward's® Breccia
Breccia Thin Section
Catalog # 470331-448
Supplier:   Avantor
Ward's® Breccia
Catalog # 470331-448
Supplier:  Avantor
Ward's® Breccia
Ward's® Breccia
Breccia Thin Section
Price
$96.59
Log in pour la disponibilité
Each
Product Details & Documents
When minerals, rocks, and fossils cannot be accurately identified by macroscopic observation and testing, thin section slides can reveal the composition and structure of the specimens on a microscopic level.
Thin section slides are precisely ground to a thickness of 0.03 mm across the entire specimen area. Mineral and rock thin sections are mounted on 27×46 mm microscope slides, and protected by a glass coverslip. All thin section slides are labeled for reference.
Spécifications
  • Désignation:
    Breccia Thin Section