Amplitude™ Sigma™ Plus Wipes, Contec
Fournisseur: Contec
New Product
Certificats
À propos de cet article
The Amplitude™ Sigma™ Plus wipes are strong and durable, compatible with most solvents, and have outstanding sorbency.
- Hydroentangled cellulose/poylester provides excellent strength and durability
- Low levels of particles and extractables
- Variety of blended fibers enable excellent absorbing capacity
Amplitude™ Sigma™ Plus Wipes are constructed using a nonwoven fabric that is a hydroentangled blend of polyester and cellulose fibers. Wet or dry, the wipe maintains a high level of strength and durability. Specially treated to reduce particles and fibers, it is recommended when low particle generation combined with sorbency is of high priority.
Certifications:
These wipes meet the requirements of USP<797> and IEST-CC-RP004.4 for 'non-shedding, low-lint, lint-free wipes'.
Frequently Bought Together