SymLine® Continuous Level Control | Avantor
SymLine® Continuous Level Control
Supplier:   SCAT AMERICAS, INC.
SymLine® Continuous Level Control
Certificates
SymLine® Continuous Level Control
SymLine® Continuous Level Control
2 Options Available Below
Product Details & Documents
Measuring filling levels continuously. Monitoring non-transparent canisters on a permanent basis is no problem with the continuous level control unit.
  • Automatic warning for 5 different filling levels freely adjustable, with accuracy of up to ±0,5 mm
  • Control box provides 5 freely configurable outlets to control external devices
  • Disc sensor for level control from non-conductive material is easy to install and adjustable in height
  • Rod sensor and canister with removable sleeve allow easy maintenance and cleaning
  • Canister made from electrostatic conductive HDPE material
The compact design and independent power supply allow simple handling and installation, also outside switch cabinets.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Capacitive rod sensor for the level measuring with ATEX-license, II 2 G EEx ia IIC T1-T6, NAMUR
    Discontinued
    Supplier Number: 108178
  • Floater, electronic, 139 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $409.31
    $409.31
    Each
       $409.31
    Supplier Number: 160123
  • Protective basket for floater, PP, suitable for all electro conductive canisters with floater
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $111.85
    $111.85
    Each
       $111.85
    Supplier Number: 199013
  • Set for continuous level control with Signalbox UM-O-Ex with 230 VAC
    Discontinued
    Supplier Number: 502042
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Built-in Signalbox2; 2-channel (EU), for SafetyWasteCap and other electronic level controlers, touchless alarm-resetting fields for 2 containers, 2 switch inputs and 2 switch outputs for peripheral units incl. 230 V power supply cable with open end and clamp
    Built-in Signalbox2; 2-channel (EU), for SafetyWasteCap and other electronic level controlers, touchless alarm-resetting fields for 2 containers, 2 switch inputs and 2 switch outputs for peripheral units incl. 230 V power supply cable with open end and clamp
    76502-952
    Log in pour la disponibilité
    Each - $1,607.97