Tyvek® 1073B Autoclave Bags without Steam Indicator, Heat Seal, Keystone Cleanroom Products
1073B Tyvek® bags are ideal for the steam sterilization of pharmaceutical filling needles.
- Manufactured and packaged in a controlled cleanroom environment
- Autoclavable at 121 to 125 °C
- Tear and puncture resistant
- Hydrophobic and lint-free
- 100% Medical Grade 1073B Tyvek® permits fast steam penetration
- Excellent impact resistance
- Consistently strong factory seals
- Exceptional microbial barrier
100% Medical Grade 1073B Tyvek® Autoclave Bags are ideal for steam sterilization of pharmaceutical filling needles and nozzless.
With consistently strong factory seals, these are a good choice for parts with sharp points.
All Tyvek construction allows for a greater number of bags per sterilization load; increasing efficiencies and cost reduction.
: Certificates of Conformance sent with each shipment.
Frequently Bought Together