J.T.Baker Tip Box for Tecan Instruments, Ritter
Fournisseur: Ritter, part of Avantor
Certificats
À propos de cet article
The J.T.Baker Tip Boxes are especially made as an accessory for pipette tips Type Tecan with C-Tray.
- Two different sizes available
Compatible tips are not included but need to be ordered separately.
Spécifications
- Catalog No:
- 45009-2000
- 45009-3000
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)