About this item
Holds large amounts of biological waste materials.
- Red in color
- Linear low density polyethylene (LLDPE)
- 1.2 mil thickness
- A high level of flexibility.
- Shear resistance can be attained without leaks
- Meets ASTM D 1709-01 and ASTM D 1922-00a specifications
For holding or consolidating larger amounts of biological waste or contaminated materials. The 33 gal. capacity bag is designed to fit Eagle 1601 series drums. Once the bag is full and ready for disposal, simply seal the bag in place and close the drum using its lid and lever-lock.
Red color bag is easily identifiable as biohazard bags, drums can be black, yellow, blue or red. Printing includes English, Spanish and French translation along with the international recognizable biohazard symbol.
Using Linear Low Density Polyethylene (LLDPE) material at 30.4 µm thickness, a high level of flexibility and shear resistance can be attained without leaks. A tubular construction, single side seal and star seal bottom provide a stable, leak proof bag.