VWR® Basic Dissecting Kit
Dissection kit
  76457-528
VWR® Basic Dissecting Kit
  76457-528
VWR® Basic Dissecting Kit
VWR® Basic Dissecting Kit
Dissection kit
$36.20
Each
Wide variety of VWR® student dissection kits that suit educational needs.
Classroom and research dissecting applications.
 : ISO, GMP
 : Includes single fold vinyl case with 6" student dissecting scalpel (standard 15" cut edge); 45" wire dissecting scissors (sharp point); 45" straight forceps (wide thumb-grip, round tip); straight teasing needle with 4" plastic handle; angular teasing needle with 4" plastic handle; glass dropper with rubber bulb (large 5" size); 6" hard plastic ruler.
  • Désignation:
    Dissection kit