VWR® Elementary Dissecting Kit - Vinyl Case
Dissection kit
  76457-526
VWR® Elementary Dissecting Kit - Vinyl Case
  76457-526
VWR® Elementary Dissecting Kit - Vinyl Case
VWR® Elementary Dissecting Kit - Vinyl Case
Dissection kit
$36.62
Each
Wide variety of VWR® student dissection kits that suit educational needs.
Classroom and research dissecting applications.
 : ISO, GMP
 : Includes single fold vinyl case with lining, 6" student dissecting scalpel (screw-lock with blade); 45" student dissecting scissors (sharp point); 45" straight forceps (wide thumb-grip, round tip); straight teasing needle with 4" plastic handle; angular teasing needle with 4" plastic handle; glass dropper with rubber bulb (large 5" size); 6" hard plastic ruler.
  • Désignation:
    Dissection kit