Corning® Organoid Counting Software for the Corning® Cell Counter, Corning
: Corning
Organoid counting software for the Corning® cell counter.
- Accurate – powered by machine learning artificial intelligence
- Multi-informative – organoids per milliliter of solution and surface area data
- Fast – three-second analysis utilizing cloud-based processing
- Cost-efficient and versatile – compatible with standard 0.1 mm counting chambers and 0.2 mm counting chambers
- No consumables required
The Organoid counting software, for use with the Corning Cell Counter, ships with a 0.2 mm, 20 µl counting chamber and an alphanumeric access code. This software extension enables Corning Cell Counter operators to capture rapid data of 3D cell cultures based on the structure’s morphology. Results are viewed within the intuitive application interface or are remotely accessible by utilizing the cloud login. Unlimited storage.
The Organoid counting software offers a size range of 20 - 200µm, a count speed of 3 seconds, and a sample volume of 10 or 20µl (chamber permitting). The field of view is 1.5×1.5mm, with a magnification of 10× and an image resolution of 1536×1536 pixels.
Frequently Bought Together