Orum International TRIO.SETTLE Table-top Holder for Petri Settling Plate, Hardy Diagnostics
Catalogue # CA76509-292
Fournisseur: HARDY DIAGNOSTICS CA
Spécifications
- Description:TRIO.SETTLE table-top holder for petri settling plate
- Cat. no.:CA76509-292
Spécifications
À propos de cet article
Benchtop format culture media plate holder with an upper surface to hold open media plate during 'passive' sampling, and a lower surface area to accommodate the lid of the media plate.
- Constructed from AISI 316 stainless steel
- Can be angled to direct the media plate’s exposure to unidirectional airflow