Desiccators, Vacuum, Clear Base, PP/PC
Desiccators, Vacuum, Clear Base, PP/PC
Desiccators, Vacuum, Clear Base, PP/PC
Desiccators, Vacuum, Clear Base, PP/PC
These autoclavable desiccators are made of polypropylene (PP) and polycarbonate (PC).
  • Can hold a vacuum up to 740 mm Hg for 24 hours without any greasing
  • Autoclavable at 120 °C for 15 minutes
  • The transparent PC bottom provides increased viewing clarity
  • An internal groove on the flange holds the silicon rubber O-ring
  • Includes a perforated PP desiccator plate
  • The knurled knob on the top allows easy handling of the dome
  • The top dome, molded in rigid and transparent PC, gives a crystal-clear view of the sample
The PP stopcock has a self-lubricating PTFE plug. The three-way plug opens to create a vacuum, closes to hold a vacuum, and opens again to release a vacuum.
Product Family Options
  • Taille
    Hauteur
    Plate Diameter
    Clearance Above Plate
    Flange I.D.
    Flange O.D.
  • 150 mm (6")
    204 mm
    138 mm
    123 mm
    150 mm
    171 mm
    Each (150mm)
    Each (150mm) - $113.45
    $113.45
    Each (150mm)
       $113.45
     : 55214
  • 200 mm (8")
    257 mm
    190 mm
    166 mm
    199 mm
    232 mm
    Each (200mm)
    Each (200mm) - $175.05
    $175.05
    Each (200mm)
       $175.05
     : 55215
  • 250 mm (10")
    315 mm
    238 mm
    207 mm
    240 mm
    276 mm
    Each (250mm)
    Each (250mm) - $217.90
    $217.90
    Each (250mm)
       $217.90
     : 55216
  • 300 mm (12")
    405 mm
    295 mm
    255 mm
    300 mm
    340 mm
    Each (300mm)
    Each (300mm) - $310.80
    $310.80
    Each (300mm)
       $310.80
     : 55217