Brady® SPC® Absorbents Chemical 14 gal. Spill Kit Refill, Brady
76518-276
- Description:Chemical spill kit refill
- Material:PP
- Cat. no.:76518-276
- Absorbency:10.1 gal.
- Details:Includes (20) 15×19" Chemical pads; (2) 3" × 12' Chemical socks; (2) disposal bags; (1) pair of goggles; (1) pair of nitrile gloves
SPC® Spill Kits from Brady® combine absorbents, accessories, PPE and instructional guidelines so yourre prepared when a spill occurs.
- Absorption capacity 10.1 gal.
- Chemical spill control
Refill kit only (container SKH-14 sold separately). Surfactant-treated, brightly colored absorbent picks up a wide range of aggressive chemicals while remaining chemically inert. Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills. Spill kits are ideal for fast response to critical accidents.