SPEC Disk SPE, Agilent Technologies
À propos de cet article
SPEC disk SPE delivers superior flow characteristics and trouble-free automation.
- No loose sorbent means no channeling and better reproducibility
- Uniform flow and extraction properties offer robust performance
- Low elution volume affords excellent concentration of the analyte, improving sensitivity
SPEC disk SPE is a product providing faster sample processing, lower costs, and high-quality results. SPEC is manufactured with ultraclean polypropylene plastic and contains a monolithic sorbent bed with bonded silica infused on a glass fiber disk. For some applications, the elution volume is very small, eliminating the dry down and reconstitution steps.
Integrated filtration protects against clogging. SPEC is available in cartridge, 96-well plate, and disk formats in a wide range of chemistries for many applications.
7 Options Available Below
Product Details & Documents
SPEC disk SPE delivers superior flow characteristics and trouble-free automation.
- No loose sorbent means no channeling and better reproducibility
- Uniform flow and extraction properties offer robust performance
- Low elution volume affords excellent concentration of the analyte, improving sensitivity
SPEC disk SPE is a product providing faster sample processing, lower costs, and high-quality results. SPEC is manufactured with ultraclean polypropylene plastic and contains a monolithic sorbent bed with bonded silica infused on a glass fiber disk. For some applications, the elution volume is very small, eliminating the dry down and reconstitution steps.
Integrated filtration protects against clogging. SPEC is available in cartridge, 96-well plate, and disk formats in a wide range of chemistries for many applications.
Product Family Options
Informations sur le produit
- DésignationType de colonnePhaseCapacitéBed WeightAvailabilityPrice
- Supplier Number: A5320320
- Supplier Number: A5322020
- Supplier Number: A532DAU
- Bond Elut cartridgeCartoucheSPEC Phenyl3 ml15 mgDiscontinued-Supplier Number: -
- Supplier Number: A5321120
- Supplier Number: A5321920
- Supplier Number: A5321130
- Bond Elut cartridgeCartoucheSPEC C18AR/MP310 ml70 mgLog in pour la disponibilitéPack of 100901,00 $Supplier Number: A5022570
Well plates
Informations sur le produit
- Catalog #DésignationVolumeAvailabilityPrice
Filter discs
Informations sur le produit
- Catalog #ØDésignationAvailabilityPrice