CrossLab Reciprocating Dissolution Screens, Agilent Technologies
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Several reciprocating vessel screens are available for the BIO-DIS reciprocating cylinder apparatus.
  • Reciprocating cylinder screens for BIO-DIS in stainless steel and polypropylene are available in different mesh sizes
Several reciprocating vessel screens are available for the Agilent BIO-DIS reciprocating cylinder apparatus, which is ideal for automatic dissolution testing of dosage forms that require different types of media.
Product Family Options
  • Désignation
  • Polypropylene screens, 0.75 inch Ext. Ø for 100 ml tubes, 40-mesh, 405 µm, for BIO-DIS, Pack of 25
    Pack of 25
    Pack of 25 - $392.00
    $392.00
    Pack of 25
       $392.00
     : 27-2205
  • Polypropylene screens, 0.75 inch Ext. Ø for 100 ml tubes, 78-mesh, 177 µm, for BIO-DIS, Pack of 25
    Pack of 25
    Pack of 25 - $392.00
    $392.00
    Pack of 25
       $392.00
     : 27-2210
  • Polypropylene screens, 0.75 inch Ext. Ø for 100 ml tubes, 100-mesh, 150 µm, for BIO-DIS, Pack of 25
    Pack of 25
    Pack of 25 - $392.00
    $392.00
    Pack of 25
       $392.00
     : 27-2215
  • Polypropylene screens, 0.75 inch Ext. Ø for 100 ml tubes, 200-mesh, 840 µm, for BIO-DIS, Pack of 25
    Pack of 25
    Pack of 25 - $392.00
    $392.00
    Pack of 25
       $392.00
     : 27-2200
  • Polypropylene screens, 1.25 inch Ext. Ø for 300 ml tubes, 20-mesh, 840 µm, for BIO-DIS, Pack of 25
    Pack of 25
    Pack of 25 - $392.00
    $392.00
    Pack of 25
       $392.00
     : 27-2000
  • Polypropylene screens, 1.25 inch Ext. Ø for 300 ml tubes, 40-mesh, 405 µm, for BIO-DIS, Pack of 25
    Pack of 25
    Pack of 25 - $392.00
    $392.00
    Pack of 25
       $392.00
     : 27-2005
  • Polypropylene screens, 1.25 inch Ext. Ø for 300 ml tubes, 56-mesh, 205 µm, for BIO-DIS, Pack of 25
    Pack of 25
    Pack of 25 - $392.00
    $392.00
    Pack of 25
       $392.00
     : 27-2007
  • Polypropylene screens, 1.25 inch Ext. Ø for 300 ml tubes, 78-mesh, 177 µm, for BIO-DIS, Pack of 25
    Pack of 25
    Pack of 25 - $392.00
    $392.00
    Pack of 25
       $392.00
     : 27-2010
  • Polypropylene screens, 1.25 inch Ext. Ø for 300 ml tubes, 100-mesh, 150 µm, for BIO-DIS, Pack of 25
    Pack of 25
    Pack of 25 - $392.00
    $392.00
    Pack of 25
       $392.00
     : 27-2015
  • Stainless Steel screens, 0.75 inch Ext. Ø for 100 ml tubes, 20-mesh, 864 µm, for BIO-DIS, Pack of 25
    Pack of 25
    Pack of 25 - $392.00
    $392.00
    Pack of 25
       $392.00
     : 27-2310
  • Stainless Steel screens, 0.75 inch Ext. Ø for 100 ml tubes, 40-mesh, 381 µm, for BIO-DIS, Pack of 25
    Pack of 25
    Pack of 25 - $392.00
    $392.00
    Pack of 25
       $392.00
     : 27-2315
  • Stainless Steel screens, 1.25 inch Ext. Ø for 300 ml tubes, 20-mesh, 864 µm, for BIO-DIS, Pack of 25
    Pack of 25
    Pack of 25 - $392.00
    $392.00
    Pack of 25
       $392.00
     : 27-2100
  • Stainless Steel screens, 1.25 inch Ext. Ø for 300 ml tubes, 30-mesh, 533 µm, for BIO-DIS, Pack of 25
    Pack of 25
    Pack of 25 - $392.00
    $392.00
    Pack of 25
       $392.00
     : 27-2103
  • Stainless Steel screens, 1.25 inch Ext. Ø for 300 ml tubes, 40-mesh, 381 µm, for BIO-DIS, Pack of 25
    Pack of 25
    Pack of 25 - $392.00
    $392.00
    Pack of 25
       $392.00
     : 27-2105
  • Stainless Steel screens, 1.25 inch Ext. Ø for 300 ml tubes, 100-mesh, 140 µm, for BIO-DIS, Pack of 25
    Pack of 25
    Pack of 25 - $392.00
    $392.00
    Pack of 25
       $392.00
     : 27-2110
  • Stainless Steel screens, 1.25 inch Ext. Ø for 300 ml tubes, 200-mesh, 74 µm, for BIO-DIS, Pack of 25
    Pack of 25
    Pack of 25 - $392.00
    $392.00
    Pack of 25
       $392.00
     : 27-2115
  • Stainless Steel screens, 1.25 inch Ext. Ø for 300 ml tubes, 300-mesh, 46 µm, for BIO-DIS, Pack of 25
    Pack of 25
    Pack of 25 - $392.00
    $392.00
    Pack of 25
       $392.00
     : 27-2120