CrossLab Dissolution Flow Cells, Agilent Technologies
CrossLab Dissolution Flow Cells, Agilent Technologies
CrossLab Dissolution Flow Cells, Agilent Technologies
CrossLab Dissolution Flow Cells, Agilent Technologies
Flow cells for the Cary UV-Vis spectrophotometers integrate dissolution testing with online UV.
  • The Cary 60 makes it easy to interconnect two different cells within the multicell transport
  • Switch between two sets of cells, typically with different path lengths, with no disassembling or reassembling of the tubing lines
Flow cells for the Agilent Cary UV-visible spectrophotometer help integrate dissolution testing with online UV analysis to provide a single-source, automated, performance-testing solution.
Product Family Options
  • Désignation
  • Flow Cell, 1 mm pathlength, for Cary 50/60
    Each
    Each - $1,860.00
    $1,860.00
    Each
       $1,860.00
     : 11-1315
  • Flow Cell, 2 mm pathlength, for Cary 50/60
    Each
    Each - $1,860.00
    $1,860.00
    Each
       $1,860.00
     : 11-1310
  • Flow Cell, 10 mm pathlength, for Cary 50/60
    Each
    Each - $1,670.00
    $1,670.00
    Each
       $1,670.00
     : 11-1300