CrossLab Dissolution Baskets, Agilent Technologies
CrossLab Dissolution Baskets, Agilent Technologies
Certificates
CrossLab Dissolution Baskets, Agilent Technologies
CrossLab Dissolution Baskets, Agilent Technologies
9 Options Available Below
Product Details & Documents
USP 40-mesh stainless steel baskets, custom baskets, and baskets for small or large dosage forms.
  • A variety of basket and mesh sizes available
  • Available in different materials and coatings
  • Individual Certificate of Conformity can be provided
From the standard USP 40-mesh stainless steel basket, to custom baskets with varying mesh sizes, to baskets specific to small or large dosage forms, Agilent supports your needs. Unless otherwise noted, all baskets fit the standard basket shafts.
 : Individual Certificate of Conformity can be provided.
Product Family Options
Informations sur le produit
  • Désignation
    Certification
    Availability
    Price
  • Basket, 100×100 Mesh, 140 µm, stainless steel, for 708-DS; 709-DS; VK7000; VK7030
    No
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,100.00
    $1,100.00
    Each
       $1,100.00
    Supplier Number: 12-2154
  • Basket, 150×150 Mesh, 104 µm, stainless steel, for 708-DS; 709-DS; VK7000; VK7030
    No
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,100.00
    $1,100.00
    Each
       $1,100.00
    Supplier Number: 12-2152
  • Basket, 270×270 Mesh, 53 µm, stainless steel, for 708-DS; 709-DS; VK7000; VK7030
    No
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,270.00
    $1,270.00
    Each
       $1,270.00
    Supplier Number: 12-2151
  • Basket, 325×2300 Mesh, 5 µm, stainless steel, for 708-DS; 709-DS; VK7000; VK7030
    No
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,270.00
    $1,270.00
    Each
       $1,270.00
    Supplier Number: 12-2146
  • Basket, 40-Mesh, 381 µm, stainless steel, with certificate, for 708-DS; 709-DS; VK7000; VK7030
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $640.00
    $640.00
    Each
       $640.00
    Supplier Number: 12-2100V
  • Basket, 50×50 Mesh, 280 µm, stainless steel, for 708-DS; 709-DS; VK7000; VK7030
    No
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $702.00
    $702.00
    Each
       $702.00
    Supplier Number: 12-2155
  • Bolus Basket, 20×20 Mesh, 864 µm, stainless steel, for 708-DS; 709-DS; VK7000; VK7030
    No
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $989.00
    $989.00
    Each
       $989.00
    Supplier Number: 12-2180
  • Bolus Basket, 40×40 Mesh, 381 µm, stainless steel, for 708-DS; 709-DS; VK7000; VK7030
    No
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $904.00
    $904.00
    Each
       $904.00
    Supplier Number: 12-2185
  • Titanium Basket kit, mini, for BIO-DIS
    No
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 1
    Kit of 1 - $2,280.00
    $2,280.00
    Kit of 1
       $2,280.00
    Supplier Number: 27-3007