Captiva Syringe Filters, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Syringe filters are often used to clarify small-volume samples before HPLC and ion chromatography.
- Ideal for busy, high-volume labs
- Available in a wide range of sizes, formats, and membranes to cover every matrix and sample type
- All Captiva premium syringe filters are shipped with an HPLC or LC/MS certificate that guarantees low levels of observed extractables
- Excellent flow rates and maximum sample loading capacities
- Every syringe filter is constructed with the highest-grade virgin polypropylene or MBS housing
- Securely welded to prevent bursting and ensure sample integrity
- Econofilters are available in money-saving packs of 1000
Captiva syringe filters offer the industry’s highest flow rates and loading capacities. Using Captiva syringe filters also significantly extends column lifetime. Choose from a variety of membrane types and pore sizes to suit your needs.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Disposable Syringes
Product Information
Frequently Bought Together