SPEC Disk SPE, Agilent Technologies | Avantor
SPEC Disk SPE, Agilent Technologies
SPEC Disk SPE, Agilent Technologies
Certificates
SPEC Disk SPE, Agilent Technologies
SPEC Disk SPE, Agilent Technologies
2 Options Available Below
Product Details & Documents
SPEC disk SPE delivers superior flow characteristics and trouble-free automation.
  • No loose sorbent means no channeling and better reproducibility
  • Uniform flow and extraction properties offer robust performance
  • Low elution volume affords excellent concentration of the analyte, improving sensitivity
SPEC disk SPE is a product providing faster sample processing, lower costs, and high-quality results. SPEC is manufactured with ultraclean polypropylene plastic and contains a monolithic sorbent bed with bonded silica infused on a glass fiber disk. For some applications, the elution volume is very small, eliminating the dry down and reconstitution steps.
Integrated filtration protects against clogging. SPEC is available in cartridge, 96-well plate, and disk formats in a wide range of chemistries for many applications.
Product Family Options
Informations sur le produit
  • Désignation
    Ø
    Availability
    Price
  • SPEC disks, C8, 47 mm
    47 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 24
    Pack of 24 - $291.00
    $291.00
    Pack of 24
       $291.00
    Supplier Number: A74702
  • SPEC disks, C18AR, 47 mm
    47 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 20
    Pack of 20 - $305.00
    $305.00
    Pack of 20
       $305.00
    Supplier Number: A74819