CrossLab Cylindrical Cells for UV-Vis and UV-Vis-NIR Systems, Agilent Technologies
Fournisseur: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificats
À propos de cet article
Agilent cylindrical cuvettes with 10 to 100 mm path length for both 8453/8454 and Cary UV systems.
- Long pathlength, 2-port cylindrical cells provide extra sensitivity for low concentration samples
- Micro cylindrical cells are for use with concentrated samples and help overcome high solvent absorbance (e.g. water in the NIR)
- Requires a cylindrical cell holder for mounting in the optical path
- Compatible with thermostatted cylindrical cell holder for added flexibility
- All cylindrical cells are optimized for use with Agilent's market-leading portfolio of UV-Vis systems
Agilent's cylindrical cuvettes (cells) enhance performance and provide added flexibility for a range of applications with a variety of path length options available. All cyclindrical cells require use of a cylindrical cell holder and are compatible with thermostatting options for flexible and accurate performance.
For low concentration samples, the 2-port, long pathlength cylindrical cells offer extra sensitivity when required for clearer spectrophotometric results every time. For concentrated samples, or to help overcome high solvent absorbance, the short pathlength cylindrical cells are recommended.
Frequently Bought Together