Cross-linked and bonded μSIL capillaries
Fournisseur: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificats
À propos de cet article
µSIL-FC and µSIL-DNA capillaries are chemically inert, hydrophobic, and stable from pH 2.5 to 10.0.
- µSIL-DNA capillaries coated with an FC polymer
- 75 µm id to accommodate the viscosity of entangled polymer solutions
- All µSIL capillaries are batch tested to ensure the highest performance and reproducibility
These capillaries are a must-have for cIEF, protein, peptide and carbohydrate separations, as well as replaceable gel CE applications such as oligonucleotides, DNA fragments, and PCR product separations. The µSIL capillaries are permanently coated using proprietary cross-linking processes to mask active silanol groups and giving high stability.
Frequently Bought Together