CrossLab Uptake PFA Tubing for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
PFA sample uptake tubing with different Int.Ø available to suit your ICP-MS application.
- PFA tubing is used to introduce sample together with the peristaltic pump tubing to the nebulizer sample inlet
- It provides chemical resistance, inertness and cleanliness
PFA tubing is used to introduce sample to the peristaltic pump tubing for the sample inlet, and to connect the outlet of this tubing to sample line connector block. This connection grounds the sample introduction tubing, eliminating any build up of electrostatic charge, that could lead to signal instability.
PFA tubing of different Int.Ø's are available so you can select the optimum tubing for your application. Tubing with a PEEK guide to ensure the sample tubing stays in the solution is also available.
Frequently Bought Together