CrossLab Spray Chamber Components for Atomic Absorption Instruments, Agilent Technologies
CrossLab Spray Chamber Components for Atomic Absorption Instruments, Agilent Technologies
CrossLab Spray Chamber Components for Atomic Absorption Instruments, Agilent Technologies
Certificates
CrossLab Spray Chamber Components for Atomic Absorption Instruments, Agilent Technologies
CrossLab Spray Chamber Components for Atomic Absorption Instruments, Agilent Technologies
12 Options Available Below
Product Details & Documents
Mark 7 atomization system for current model Agilent AA instruments.
  • Simple-to-use twist and lock assembly makes for easy cleaning and routine maintenance
  • Flexible operation with the capability to tune performance using the externally adjustable impact bead, fit mixing paddles for better precision, or both
  • Inert construction makes spray chamber suitable for use with aqueous and organic solutions
  • Fit organic resistant O-rings to ensure organic compatibility
The nebulizer, spray chamber and burner, plays an important part in controlling the analytical performance of a flame AA spectrometer. Proper spray chamber design ensures the minimization of carbon build-up, burner blockage and volatilization interferences. The spray chamber must also provide excellent drainage.
The Mark 7 spray chamber features an adjustable impact bead that breaks up aerosol droplets and provides the operator with control over the aerosol concentration. A set of removable twin-head mixing paddles promote mixing and large droplet removal. These features provide a smaller drop size aerosol, reducing burner blockage, signal noise, and improving long term signal stability.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • 6210092090 - Moulding Spraychamber Fluorinated
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 215,00 $
    215,00 $
    Each
       215,00 $
    Supplier Number: 6210092090
  • Blanking plug, nebulizer block, PEEK, for Mark 7 spray chamber
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 28,30 $
    28,30 $
    Each
       28,30 $
    Supplier Number: 1610118400
  • Impact bead securing screw, PEEK, M6 × 6LG, for Mark 7 spray chamber
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 21,00 $
    21,00 $
    Each
       21,00 $
    Supplier Number: 1510226500
  • Kit, fluorinated liquid trap, float and magnet
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 1
    Kit of 1 - 546,00 $
    546,00 $
    Kit of 1
       546,00 $
    Supplier Number: 9910105800
  • Mixing paddle, fluorinated, Mark 7, for Mark 7 spray chamber
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 74,90 $
    74,90 $
    Each
       74,90 $
    Supplier Number: 110637000
  • Mixing paddles
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 5
    Pack of 5 - 392,00 $
    392,00 $
    Pack of 5
       392,00 $
    Supplier Number: 9910063600
  • Mixing paddles, fluorinated
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 5
    Pack of 5 - 323,00 $
    323,00 $
    Pack of 5
       323,00 $
    Supplier Number: 9910093600
  • Nebulizer block, fluorinated, for Mark 7 spray chamber, excludes integrated nebulizer
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 750,00 $
    1 750,00 $
    Each
       1 750,00 $
    Supplier Number: 9910093100
  • Screw, bead adjuster, gray, PEEK, for Mark 7 spray chamber
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 55,20 $
    55,20 $
    Each
       55,20 $
    Supplier Number: 1510225000
  • Screw, spray chamber mounting, for Mark 7 spray chamber
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 48,40 $
    48,40 $
    Each
       48,40 $
    Supplier Number: 1510226600
  • Screw, stainless steel, bead adjuster, locking, M8, for Mark 7 spray chamber
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 37,10 $
    37,10 $
    Each
       37,10 $
    Supplier Number: 1510226000
  • Spray chamber bung, fluorinated
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 106,00 $
    106,00 $
    Each
       106,00 $
    Supplier Number: 1610118800