CrossLab Gas Chromatography (GC) Interface Supplies for ICP-MS
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
New Product
Certificates
About this item
Agilent is an expert in speciation analysis, offering GC-ICP-MS parts and interface supplies.
- The Agilent GC-ICP-MS interface consists of a heated, passivated transfer line and a special torch with a heated injector tube
- The unique interface is ideal for routine analysis of high-boiling point compounds, providing unparalleled performance
- All GC-ICP-MS interface supplies are designed to Agilent’s demanding specifications and can help to achieve the market-leading ICP-MS performance that our valued clients have come to expect
- The GC interface function is enabled with PFA tubes, transfer lines and torches for the specific interface along with stainless pipe (Sulfinert) and clamp plates (PEEK) for straightforward installation and maintenance
- Argon make-up gas supplied by the ICP-MS is preheated via a stainless steel heat exchanger inside the GC oven before being added to the GC column outlet to maintain constant high temperature from the end of the GC column to the tip of the ICP injector
The Agilent GC-ICP-MS interface consists of a heated, passivated transfer line and a special torch with a heated injector tube. The unique GC-ICP-MS interface is ideal for routine analysis of high-boiling point compounds, providing unparalleled performance for advanced GC-ICP-MS applications.
Argon make-up gas supplied by the ICP-MS is preheated via a stainless steel heat exchanger inside the GC oven before being added to the GC column effluent at the head of the transfer line. In this way, the sample is maintained at constant high temperature from the end of the chromatographic column in the GC oven to the tip of the ICP injector.
Frequently Bought Together