Filtering Flasks, with Plastic Hose Connection, Sartorius
Fournisseur: Sartorius
Certificats
À propos de cet article
These vacuum-resistant filtering flasks made of DURAN® borosilicate glass, can be used in filtrate collection.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Standard according to DIN 12476.
Frequently Bought Together