VF-1ms Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
A highly inert, nonpolar, low-bleed, PDMS phase, ideal for GC/MS and for trace analysis in a wide range of applications.
- Highly inert, nonpolar, 100% dimethylpolysiloxane phase
- Low-bleed GC column providing increased sensitivity over a broad range of applications
- QC test results for retention index, efficiency, selectivity, and bleed reported with every column
- 0.15 mm Int.Ø columns available for high efficiency GC and GC/MS analysis
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
Agilent J&W VF-1ms is a highly inert, nonpolar, 100% dimethylpolysiloxane phase column that provides increased sensitivity for a broad array of applications, particularly for energy, pharmaceutical, and environmental analysis. The column has an ultra-low-bleed specification of 1 pA at 325 °C (30 m, 0.25 mm, 0.25 μm) for trace analysis with GC/MS.
Specifications
- Catalog No:
- CAAGCP8916
- CAAGCP8913
- CAAGCP8907
- CAAGCP8912
- CAAGCP8904
- CAAGCP8930
- Polarity:
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase