J&W PoraPLOT Q Columns, Agilent Technologies
À propos de cet article
A porous polymer PLOT stationary phase for analysis of polar and apolar volatile compounds, and water as an analyte.
- Long-term stability provides repeatable retention times
- Available in Fused Silica and UltiMetal
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
- Similar phases: Rt-Q BOND, Rt-QPLOT, SupelQ PLOT
Agilent J&W PoraPLOT Q is recommended for column switching systems that analyze polar and apolar volatile compounds. Water elutes as a sharp and quantifiable peak, making retention times of target compounds repeatable, as they are not influenced by water in the sample.
Columns with 7" cage.
8 Options Available Below
Product Details & Documents
A porous polymer PLOT stationary phase for analysis of polar and apolar volatile compounds, and water as an analyte.
- Long-term stability provides repeatable retention times
- Available in Fused Silica and UltiMetal
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
- Similar phases: Rt-Q BOND, Rt-QPLOT, SupelQ PLOT
Agilent J&W PoraPLOT Q is recommended for column switching systems that analyze polar and apolar volatile compounds. Water elutes as a sharp and quantifiable peak, making retention times of target compounds repeatable, as they are not influenced by water in the sample.
Columns with 7" cage.
Product Family Options
Informations sur le produit
- DésignationType de colonnePhaseColumn HardwareÉpaisseur du filmØ int.LongueurAvailabilityPrice
- GC column, 5" cageCapillairePoraPLOT QFused silica10.00 µm0.32 mm10 mLog in pour la disponibilitéEach880,00 $Supplier Number: CP7550I5
- GC column, 7" cageCapillairePoraPLOT QFused silica8.00 µm0.25 mm25 mLog in pour la disponibilitéEach1 060,00 $Supplier Number: CP7549
- GC column, 7" cageCapillairePoraPLOT QFused silica10.00 µm0.32 mm10 mLog in pour la disponibilitéEach785,00 $Supplier Number: CP7550
- GC column, 7" cageCapillairePoraPLOT QFused silica10.00 µm0.32 mm25 mLog in pour la disponibilitéEach1 140,00 $Supplier Number: CP7551
- GC column, 7" cageCapillairePoraPLOT QFused silica20.00 µm0.53 mm25 mLog in pour la disponibilitéEach1 230,00 $Supplier Number: CP7554
- GC column, 7" cageCapillairePoraPLOT QFused silica20.00 µm0.53 mm10 mLog in pour la disponibilitéEach776,00 $Supplier Number: CP7553
- GC column, 7" cageCapillairePoraPLOT QFused silica10.00 µm0.32 mm50 mLog in pour la disponibilitéEach1 730,00 $Supplier Number: CP7552
- GC column, 7" cageCapillairePoraPLOT QAcier inoxydable20.00 µm0.53 mm25 mLog in pour la disponibilitéEach1 460,00 $Supplier Number: CP6954