About this item
Designed for the analysis of permanent gases.
- A PLOT column for the analysis of permanent gases
- O₂, N₂, CO, and CH₄ resolve in less than 5 minutes
- Durable molecular sieve 5Å coating minimizes baseline spiking and damage to multiport valves
- Select a thick film for Ar/O₂ separation without cryogenic cooling
- Select thin film HP-PLOT Molesieve columns for routine air monitoring applications
- Replaces GS-Molesieve
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each GC column
Note: Molecular sieve columns absorb water, which over time results in changes in retention time. We use an advanced, proprietary deactivation process that allows for rapid regeneration. Fully saturated HP-PLOT Molesieve columns regenerate in 7 hours or less at 200 °C
Agilent J&W HP-PLOT Molesieve are offered in 0.53 and 0.32 mm Int. Ø. For applications requiring resolution of argon and oxygen without expensive cryogenic cooling, select thick-film HP-PLOT Molesieve columns. The thin-film configurations are ideal for many applications, including routine air monitoring and analysis in less than 10 seconds. The thin-film column resolves argon and oxygen at subambient temperatures.
All columns are with 7" cage.