J&W HP-FFAP Columns, Agilent Technologies
J&W HP-FFAP Columns, Agilent Technologies
Certificates
J&W HP-FFAP Columns, Agilent Technologies
13 Options Available Below
Product Details & Documents
HP-FFAP is a high-polarity phase designed for the analysis of volatile fatty acids and phenols.
  • Nitroterephthalic acid modified polyethylene glycol
  • Designed for the analysis of volatile fatty acids and phenols
  • Replaces OV-351
  • Bonded and cross-linked, solvent rinsable
  • Close equivalent to USP Phase G35
  • Similar Phases: Stabilwax-DA, Nukol, 007-FFAP, BP21, AT-1000, OV-351
Agilent J&W HP-FFAP is designed primarily for the analysis of organic acids, free fatty acids, or samples that require quantitation of acidic impurities. The FFAP stationary phase is modified with acid to provide a very inert column that can accommodate the demanding analysis of acids dissolved in water.;Free fatty acids up to C24 can be analyzed by HP-FFAP without costly and time-consuming derivatization. HP-FFAP does not require priming (several injections of acids), so accurate results can be obtained quickly.
Each Agilent J&W GC column is tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency to provide you utmost confidence in qualitative and quantitative results. For details about the QC test conditions including the test probe mixture and the resulting chromatogram, please refer to the Agilent J&W GC Column Performance Summary shipped with each GC column.
Note: We do not recommend the use of water or methanol to rinse HP-FFAP GC columns.
All columns are with 7" cage.
Specifications for Product Family
Spécifications
  • Catalog No:
  • CA19091F-102
  • CAAG19091F102E
  • CAAG19091F-105
  • CA19091F-433
  • CA19091F-112
  • CAAG19091F112E
  • CA19091F-413
  • CAAG19091F115E
  • CAAG19091F-115
  • CAAG19095F-121
  • CA19095F-120
  • CA19095F-123
  • CAAG19095F123E
  • Polarity:
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Type de colonne
    Phase
    Column Hardware
    Épaisseur du film
    Ø int.
    Longueur
    Availability
    Price
  • Capillaire
    HP-FFAP
    Fused silica
    0.33 µm
    0.20 mm
    25 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $830.00
    $830.00
    Each
       $830.00
    Supplier Number: 19091F-102
    Spécifications:
    More
    Specifications
    Cat. No.
    CA19091F-102
    Polarity
    High-polarity stationary phase
  • GC column, 5" cage
    Capillaire
    HP-FFAP
    Fused silica
    0.33 µm
    0.20 mm
    25 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,030.00
    $1,030.00
    Each
       $1,030.00
    Supplier Number: 19091F-102E
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG19091F102E
    Polarity
    High-polarity stationary phase
  • GC column, 7" cage
    Capillaire
    HP-FFAP
    Fused silica
    0.33 µm
    0.20 mm
    50 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,630.00
    $1,630.00
    Each
       $1,630.00
    Supplier Number: 19091F-105
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG19091F-105
    Polarity
    High-polarity stationary phase
  • Capillaire
    HP-FFAP
    Fused silica
    0.25 µm
    0.25 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $983.00
    $983.00
    Each
       $983.00
    Supplier Number: 19091F-433
    Spécifications:
    More
    Specifications
    Cat. No.
    CA19091F-433
    Polarity
    High-polarity stationary phase
  • Capillaire
    HP-FFAP
    Fused silica
    0.50 µm
    0.32 mm
    25 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $941.00
    $941.00
    Each
       $941.00
    Supplier Number: 19091F-112
    Spécifications:
    More
    Specifications
    Cat. No.
    CA19091F-112
    Polarity
    High-polarity stationary phase
  • GC column, 5" cage
    Capillaire
    HP-FFAP
    Fused silica
    0.50 µm
    0.32 mm
    25 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,170.00
    $1,170.00
    Each
       $1,170.00
    Supplier Number: 19091F-112E
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG19091F112E
    Polarity
    High-polarity stationary phase
  • Capillaire
    HP-FFAP
    Fused silica
    0.25 µm
    0.32 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,070.00
    $1,070.00
    Each
       $1,070.00
    Supplier Number: 19091F-413
    Spécifications:
    More
    Specifications
    Cat. No.
    CA19091F-413
    Polarity
    High-polarity stationary phase
  • GC column, 5" cage
    Capillaire
    HP-FFAP
    Fused silica
    0.50 µm
    0.32 mm
    50 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,080.00
    $2,080.00
    Each
       $2,080.00
    Supplier Number: 19091F-115E
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG19091F115E
    Polarity
    High-polarity stationary phase
  • GC column, 7" cage
    Capillaire
    HP-FFAP
    Fused silica
    0.50 µm
    0.32 mm
    50 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,860.00
    $1,860.00
    Each
       $1,860.00
    Supplier Number: 19091F-115
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG19091F-115
    Polarity
    High-polarity stationary phase
  • GC column, 7" cage
    Capillaire
    HP-FFAP
    Fused silica
    1.00 µm
    0.53 mm
    10 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $593.00
    $593.00
    Each
       $593.00
    Supplier Number: 19095F-121
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG19095F-121
    Polarity
    High-polarity stationary phase
  • Capillaire
    HP-FFAP
    Fused silica
    1.00 µm
    0.53 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $683.00
    $683.00
    Each
       $683.00
    Supplier Number: 19095F-120
    Spécifications:
    More
    Specifications
    Cat. No.
    CA19095F-120
    Polarity
    High-polarity stationary phase
  • Capillaire
    HP-FFAP
    Fused silica
    1.00 µm
    0.53 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,180.00
    $1,180.00
    Each
       $1,180.00
    Supplier Number: 19095F-123
    Spécifications:
    More
    Specifications
    Cat. No.
    CA19095F-123
    Polarity
    High-polarity stationary phase
  • GC column, 5" cage
    Capillaire
    HP-FFAP
    Fused silica
    1.00 µm
    0.53 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,460.00
    $1,460.00
    Each
       $1,460.00
    Supplier Number: 19095F-123E
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG19095F123E
    Polarity
    High-polarity stationary phase