HP-5ms Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Nonpolar, low-bleed columns with excellent inertness for active compounds, including acidic and basic compounds.
- (5%-Phenyl)-methylpolysiloxane
- Identical selectivity to HP-5
- Nonpolar with very low bleed characteristics, ideal for GC/MS
- Excellent inertness for active compounds, including acidic and basic compounds
- Improved signal-to-noise ratio for better sensitivity and mass spectral integrity
- Bonded, crosslinked, and solvent rinsable
- Equivalent to USP phase G27
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
- Similar phases: Rtx-5ms, Rxi-5ms, Rxi-5Sil MS, PTE-5, BPX-5, AT-5ms, ZB-5ms, ZB-5ms Plus, SLB-5ms, Equity-5
Agilent J&W HP-5ms is a (5%-phenyl)-methylpolysiloxane phase with very low bleed characteristics that is ideal for GC/MS. The column is bonded, crosslinked, and solvent rinsible, and has excellent inertness for active compounds, including acidic and basic compounds, with improved signal-to-noise ratio for better sensitivity and mass spectral integrity.
Equivalent to USP phase G27, it delivers excellent performance across a range of applications.
Specifications
- Catalog No:
- CA19091S-413
- CAAG19091S433I
- CAAG19091S577U
- CAAG19091S233I
- CA19091S-436
- CA19091S-102
- CA19091S-101
- CA19091S-431
- CA19091S-433E
- CAAG19091S111L
- CA76514-574
- CAAG19091S-113
- CAAG19091S-133
- CAAG19091S-105
- CAAG19091S-112
- CAAG19091S-577
- CAAG19091S577N
- CA19091S-139
- CAAG19091S433N
- CAAG19091S-336
- CAAG19091S-416
- CAAG19091S-231
- CAAG19091S-233
- CAAG19091S-331
- CAAG19091S-333
- CAAG19091S-213
- Polarity:
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase