HP-5 Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
An excellent nonpolar, general-purpose 5% phenyl GC column with high temperature limits for a wide range of applications.
- (5%-Phenyl)-methylpolysiloxane
- An excellent nonpolar, general-purpose column for a wide range of applications
- High temperature limit
- Bonded, crosslinked, and solvent rinsible
- Equivalent to USP phase G27
- Wide range of column dimensions available
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
Agilent J&W HP-5 is a precision-engineered (5%-phenyl)-methylpolysiloxane nonpolar column that delivers superb performance in a wide range of applications. This general-purpose column is bonded, crosslinked, and solvent rinsible and is equivalent to USP phase G27.
Specifications
- Catalog No:
- CA19091J-215
- CA19095J-623
- CAAG19091J215E
- CAAG19091J216E
- CAAG19091J-005
- CA19095J-321
- CAAG19091J-436
- CAAG19091J-577
- CAAG19091J012E
- CAAG19091J112E
- CAAG19091J-236
- CAAG19095J-323
- CAAG19091J-333
- CAAG19095J-621
- CAAG19091J-411
- CA76514-570
- CAAG19091J-416
- CAAG19095J-123
- CA19095J-121
- CA19091J-413E
- CA19091J-012
- CA19091J-202
- CAAG19091J-216
- CA19091J-433
- CAAG19095J-023
- CAAG19091J-231
- CA19091J-431
- CA19091J-101
- CAAG19091J-233
- CAAG19091J-115
- CAAG19091J-205
- CAAG19091J-212
- CAAG19091J-213
- CAAG19091J413I
- CAAG19091J433E
- CAAG19091J-102
- CAAG19091J577L
- CAAG19091J-112
- CAAG19091J-113
- CAAG19091J411E
- Polarity:
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase