J&W HP-PLOT Al2O3 S Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
A deactivated, mid-polarity PLOT column for light hydrocarbon analysis, particularly C1 to C8 hydrocarbon isomers.
- Middle range of polarity for alumina phases
- Aluminum oxide deactivated with sodium sulfate
- Excellent general-use column for light hydrocarbon analysis (C1–C8 isomers)
- Best for resolving acetylene from butane and propylene from isobutane
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
Agilent J&W HP-PLOT Al₂O₃ S is the standard column choice for light hydrocarbon analysis, particularly C1 to C8 hydrocarbon isomers. A mid-polar range alumina phase deactivated with sodium sulfate, it is excellent for resolving acetylene from butylene and isobutane.
Frequently Bought Together