GS-GasPro Columns, Agilent Technologies
A unique bonded silica PLOT column that is an excellent choice for light hydrocarbons and sulfur gases.
- Unique bonded PLOT column technology
- Excellent choice for light hydrocarbons and sulfur gases
- Retention stability not affected by water
- Separates CO and CO₂ on a single column
- No particles, making it an ideal PLOT column for GC/MS
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
- Similar phases: CP-Silica PLOT
The Agilent J&W GS-GasPro column is precision-engineered with unique bonded silica PLOT column technology, making it an excellent choice for light hydrocarbons and sulfur gases. With an operating temperature range of –80 to 260/300 °C and extensive testing to strict industry QC standards, it delivers the level of performance demanded by analytical GC/MS.
The column features high retention stability that is unaffected by water and enables separation of CO and CO2 on a single column.
Frequently Bought Together