DB-5ms Columns, Agilent Technologies
DB-5ms Columns, Agilent Technologies
DB-5ms Columns, Agilent Technologies
Certificates
DB-5ms Columns, Agilent Technologies
39 Options Available Below
Product Details & Documents
A nonpolar column with high inertness and improved signal-to-noise for analysis of aromatics and semi volatiles.
  • Phenyl arylene polymer virtually equivalent to (5%-phenyl)-methylpolysiloxane
  • Nonpolar with very low bleed characteristics, ideal for GC/MS
  • Excellent inertness for active compounds
  • Improved signal-to-noise ratio for better sensitivity and mass spectral integrity
  • Bonded, crosslinked, and solvent rinsible
  • Exact replacement of HP-5TA
  • Close equivalent to USP phase G27
  • Test mix available
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency Performance Summary shipped with each column
Agilent J&W DB-5ms is a nonpolar phenyl arylene polymer, virtually equivalent to (5%-phenyl)-methylpolysiloxane, featuring an excellent signal-to-noise ratio for analytical applications. This improved performance gives better sensitivity and mass spectral integrity and delivers excellent inertness with active compounds.
This column is a close equivalent to USP phase G27, and is very low bleed, making it ideal for GC/MS.
Specifications for Product Family
Spécifications
  • Catalog No:
  • CA121-5522
  • CA122-5513
  • CA122-5516
  • CA122-5531
  • CA122-5533
  • CA122-5536
  • CA122-5561
  • CA122-5562
  • CA123-5532
  • CA123-5533
  • CA123-5536
  • CA123-5563
  • CA125-5532
  • CA128-5512
  • CA128-5522
  • CA76514-542
  • CA76514-546
  • CAAG122-5532E
  • CAAG122-5511
  • CAAG122-5522
  • CAAG122-5533E
  • CAAG122-5552
  • CAAG122-5562E
  • CAAG122-5563
  • CAAG1225512LTM
  • CAAG1225516INT
  • CAAG1225532INT
  • CAAG123-5511
  • CAAG123-5512
  • CAAG123-5513
  • CAAG123-5526
  • CAAG123-5531
  • CAAG123-5532E
  • CAAG123-5562
  • CAAG123-5566
  • CAAG125-5512
  • CAAG125-5537
  • CAAG125-553J
  • CAAG128-5552
  • Polarity:
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • USP code:
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Type de colonne
    Phase
    Column Hardware
    Épaisseur du film
    Ø int.
    Longueur
    Availability
    Price
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    0.18 µm
    0.18 mm
    20 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $901.00
    $901.00
    Each
       $901.00
    Supplier Number: 121-5522
    Spécifications:
    More
    Specifications
    Cat. No.
    CA121-5522
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    1.00 µm
    0.25 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $645.00
    $645.00
    Each
       $645.00
    Supplier Number: 122-5513
    Spécifications:
    More
    Specifications
    Cat. No.
    CA122-5513
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    0.50 µm
    0.25 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $645.00
    $645.00
    Each
       $645.00
    Supplier Number: 122-5516
    Spécifications:
    More
    Specifications
    Cat. No.
    CA122-5516
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    0.10 µm
    0.25 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,060.00
    $1,060.00
    Each
       $1,060.00
    Supplier Number: 122-5531
    Spécifications:
    More
    Specifications
    Cat. No.
    CA122-5531
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    1.00 µm
    0.25 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,060.00
    $1,060.00
    Each
       $1,060.00
    Supplier Number: 122-5533
    Spécifications:
    More
    Specifications
    Cat. No.
    CA122-5533
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    0.50 µm
    0.25 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,060.00
    $1,060.00
    Each
       $1,060.00
    Supplier Number: 122-5536
    Spécifications:
    More
    Specifications
    Cat. No.
    CA122-5536
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    0.10 µm
    0.25 mm
    60 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,800.00
    $1,800.00
    Each
       $1,800.00
    Supplier Number: 122-5561
    Spécifications:
    More
    Specifications
    Cat. No.
    CA122-5561
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    0.25 µm
    0.25 mm
    60 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,800.00
    $1,800.00
    Each
       $1,800.00
    Supplier Number: 122-5562
    Spécifications:
    More
    Specifications
    Cat. No.
    CA122-5562
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    0.25 µm
    0.32 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,150.00
    $1,150.00
    Each
       $1,150.00
    Supplier Number: 123-5532
    Spécifications:
    More
    Specifications
    Cat. No.
    CA123-5532
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    1.00 µm
    0.32 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,150.00
    $1,150.00
    Each
       $1,150.00
    Supplier Number: 123-5533
    Spécifications:
    More
    Specifications
    Cat. No.
    CA123-5533
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    0.50 µm
    0.32 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,150.00
    $1,150.00
    Each
       $1,150.00
    Supplier Number: 123-5536
    Spécifications:
    More
    Specifications
    Cat. No.
    CA123-5536
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    1.00 µm
    0.32 mm
    60 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,960.00
    $1,960.00
    Each
       $1,960.00
    Supplier Number: 123-5563
    Spécifications:
    More
    Specifications
    Cat. No.
    CA123-5563
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    1.50 µm
    0.53 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,260.00
    $1,260.00
    Each
       $1,260.00
    Supplier Number: 125-5532
    Spécifications:
    More
    Specifications
    Cat. No.
    CA125-5532
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    0.33 µm
    0.20 mm
    12 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $570.00
    $570.00
    Each
       $570.00
    Supplier Number: 128-5512
    Spécifications:
    More
    Specifications
    Cat. No.
    CA128-5512
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    0.33 µm
    0.20 mm
    25 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $875.00
    $875.00
    Each
       $875.00
    Supplier Number: 128-5522
    Spécifications:
    More
    Specifications
    Cat. No.
    CA128-5522
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    0.25 µm
    0.25 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $710.00
    $710.00
    Each
       $710.00
    Supplier Number: 122-5512
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76514-542
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    0.25 µm
    0.25 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,170.00
    $1,170.00
    Each
       $1,170.00
    Supplier Number: 122-5532
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76514-546
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 5" cage
    Capillaire
    DB-5ms
    Fused silica
    0.25 µm
    0.25 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,310.00
    $1,310.00
    Each
       $1,310.00
    Supplier Number: 122-5532E
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG122-5532E
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    0.1 µm
    0.25 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $710.00
    $710.00
    Each
       $710.00
    Supplier Number: 122-5511
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG122-5511
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    0.25 µm
    0.25 mm
    25 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,020.00
    $1,020.00
    Each
       $1,020.00
    Supplier Number: 122-5522
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG122-5522
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 5" cage
    Capillaire
    DB-5ms
    Fused silica
    1.00 µm
    0.25 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,310.00
    $1,310.00
    Each
       $1,310.00
    Supplier Number: 122-5533E
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG122-5533E
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    0.25 µm
    0.25 mm
    50 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,820.00
    $1,820.00
    Each
       $1,820.00
    Supplier Number: 122-5552
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG122-5552
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 5" cage
    Capillaire
    DB-5ms
    Fused silica
    0.25 µm
    0.25 mm
    60 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,230.00
    $2,230.00
    Each
       $2,230.00
    Supplier Number: 122-5562E
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG122-5562E
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    1.00 µm
    0.25 mm
    60 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,980.00
    $1,980.00
    Each
       $1,980.00
    Supplier Number: 122-5563
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG122-5563
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • LTM column module
    Capillaire
    DB-5ms
    Fused silica
    0.25 µm
    0.25 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,250.00
    $2,250.00
    Each
       $2,250.00
    Supplier Number: 122-5512LTM
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG1225512LTM
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column module
    Capillaire
    DB-5ms
    Fused silica
    0.50 µm
    0.25 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $824.00
    $824.00
    Each
       $824.00
    Supplier Number: 122-5516-INT
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG1225516INT
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column module
    Capillaire
    DB-5ms
    Fused silica
    0.25 µm
    0.25 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,350.00
    $1,350.00
    Each
       $1,350.00
    Supplier Number: 122-5532-INT
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG1225532INT
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    0.1 µm
    0.32 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $767.00
    $767.00
    Each
       $767.00
    Supplier Number: 123-5511
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG123-5511
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    0.25 µm
    0.32 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $767.00
    $767.00
    Each
       $767.00
    Supplier Number: 123-5512
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG123-5512
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    1.00 µm
    0.32 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $767.00
    $767.00
    Each
       $767.00
    Supplier Number: 123-5513
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG123-5513
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    0.52 µm
    0.32 mm
    25 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,080.00
    $1,080.00
    Each
       $1,080.00
    Supplier Number: 123-5526
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG123-5526
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    0.1 µm
    0.32 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,260.00
    $1,260.00
    Each
       $1,260.00
    Supplier Number: 123-5531
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG123-5531
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 5" cage
    Capillaire
    DB-5ms
    Fused silica
    0.25 µm
    0.32 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,410.00
    $1,410.00
    Each
       $1,410.00
    Supplier Number: 123-5532E
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG123-5532E
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    0.25 µm
    0.32 mm
    60 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,160.00
    $2,160.00
    Each
       $2,160.00
    Supplier Number: 123-5562
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG123-5562
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    0.50 µm
    0.32 mm
    60 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,160.00
    $2,160.00
    Each
       $2,160.00
    Supplier Number: 123-5566
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG123-5566
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    1.50 µm
    0.53 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $787.00
    $787.00
    Each
       $787.00
    Supplier Number: 125-5512
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG125-5512
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    0.50 µm
    0.53 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,380.00
    $1,380.00
    Each
       $1,380.00
    Supplier Number: 125-5537
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG125-5537
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    1.00 µm
    0.53 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,380.00
    $1,380.00
    Each
       $1,380.00
    Supplier Number: 125-553J
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG125-553J
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5ms
    Fused silica
    0.33 µm
    0.20 mm
    50 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,630.00
    $1,630.00
    Each
       $1,630.00
    Supplier Number: 128-5552
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG128-5552
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41