DB-35ms Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Virtually equivalent to a (35%-phenyl) methylpolysiloxane, with excellent inertness and low bleed, ideal for confirmation analysis.
- Virtually equivalent to (35%-phenyl)-methylpolysiloxane
- Mid polarity with very low bleed characteristics, ideal for GC/MS
- Extended temperature limit of 340/360 °C
- Excellent inertness for active compounds
- Ideal for confirmation analyses
- Close equivalent to USP phase G42
- Bonded, crosslinked, and solvent rinsible
- Replaces HP-35ms
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
Agilent J&W DB-35ms, a close equivalent to USP phase G42, is precision engineered to deliver excellent performance in a range of GC/MS applications, particularly in confirmation analysis. These bonded, crosslinked, mid-polarity columns operate over an extended temperature range up to 340/360 °C. They feature very low-bleed characteristics and excellent inertness and are particularly well suited for analysis of active compounds.
Specifications
- Catalog No:
- CA125-3832
- CA122-3812
- CA123-3832
- CA122-3832
- CA128-3812
- CAAG122-3862
- CAAG125-3837
- CAAG128-3822
- CAAG122-3831
- CAAG1283822INT
- Polarity:
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- USP code:
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42