J&S DB-210 Columns, Agilent Technologies
Fournisseur: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificats
À propos de cet article
Excellent for EPA methods 8140 and 609 and is high polarity, bonded, crosslinked, and solvent rinsable.
- (50%-Trifluoropropyl)-methylpolysiloxane
- Excellent for US EPA Methods 8140 and 609
- High polarity, bonded, crosslinked, and solvent rinsable
- Exact replacement of HP-210
- Close equivalent to USP Phase G6
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
Agilent J&W DB-210 is particularly designed for optimized for EPA methods 8140 and 609. Bonded and crosslinked, these columns are close equivalents to USP phase G6 and are efficient and solvent rinsable. With effective operation between 45 °C and 240/260 °C, and strict QC testing to the highest industry standards, DB-210 columns provide robust, accurate, and reproducible performance for a wide range of GC/MS applications.
Spécifications
- Catalog No:
- CA123-0233
- CAAG125-0212
- CAAG123-0232
- CAAG123-0213
- CA122-0233
- CAAG125-0232
- Polarity:
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- USP code:
- G6
- G6
- G6
- G6
- G6
- G6