J&W DB-17ht Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
A high-temperature GC column for improved resolution of triglycerides, ideal for confirmation analysis at elevated temperature.
- (50%-Phenyl)-methylpolysiloxane
- Mid-polarity
- Extended upper temperature limit of 365 °C
- High temperature, polyimide-coated, fused silica tubing
- Excellent peak shape and faster elution times for high boilers
- Improved resolution for triglycerides
- Ideal for confirmation analysis
- Bonded, crosslinked, and solvent rinsable
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
- Similar phases: Rtx-65TG, BPX50
Agilent J&W DB-17ht provides excellent performance for accurate extended temperature range analysis. These mid-polarity columns are solvent rinsable and feature a special high-temperature polyimide-coated fused silica tubing. Stable and efficient manufacturing, utilizing high-quality components and processes, results in improved resolution for the effective analysis of triglycerides and other high boilers over extended temperature limits of 40 to 340/365 °C.
Specifications
- Catalog No:
- CA122-1831
- CAAG122-1811
- CAAG122-1801
- CAAG123-1831
- Polarity:
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase