J&W DB-1701 Columns, Agilent Technologies
A low/mid polarity column ideal for the analysis of pesticides, herbicides, TMS sugars, and aroclors.
- (14%-Cyanopropyl-phenyl)-methylpolysiloxane
- Solvent rinsable with low/mid polarity
- Bonded and crosslinked
- Exact replacement of HP-1301 and HP-1701
- Available with DuraGuard (the GC column with a built-in guard column)
- Tested against the strictest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
- Similar phases: SPB-1701, Rtx-1701, BP-10, OV-1701, 007-1701, ZB-1701
Agilent J&W DB-1701 is a low/mid-polarity (14% cyanopropyl-phenyl)-methylpolysiloxane column. It is ideal for the analysis of pesticides, herbicides, trimethylsilyl (TMS) derivatives of sugars and related substances, and aroclors. The column is bonded, crosslinked, and solvent rinsable and is available with DuraGuard (the GC column with a built-in guard column).
- Catalog No:
- CA122-0733
- CA123-0712
- CA122-0732
- CA123-0763
- CA123-0732
- CA122-0762
- CAAG125-0762
- CAAG127-0722
- CAAG125-0733
- CAAG125-0731
- CAAG127-0723
- CAAG123-0733E
- CA125-0732
- CAAG123-0732E
- CAAG123-0731
- CA125-0712
- CA123-0733
- CAAG123-0713
- CAAG1230733INT
- CAAG123-0762
- CAAG121-0722
- CAAG123-0753
- Polarity:
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- USP code:
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
Frequently Bought Together