CP-Sil 5 CB Columns, Agilent Technologies
A low-polarity, general-purpose, 100% dimethylpolysiloxane (PDMS) column ideal for a wide range of applications.
- 100% dimethylpolysiloxane (PDMS)
- Nonpolar
- General purpose phase
- Bonded, crosslinked, and solvent rinsible
- Separates almost entirely based on boiling points, making this column suitable for a wide range of applications with a broad temperature range
- Available in Fused Silica or UltiMetal
- Tested against the strictest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
- Similar phases: SPB-1, Rtx-1, BP-1, OV-1, OV-101, 007-1(MS), SP-2100, SE-30, ZB-1, AT-1, ZB-1
Agilent J&W CP-Sil 5 CB is a general-purpose, nonpolar column containing a 100% dimethylpolysiloxane (PDMS) phase. Separation is almost entirely based on boiling points, making this column suitable for a wide range of applications with a broad temperature range.
Due to extensive crosslinking, CP-Sil 5 CB is highly inert and withstands large solvent injections, guaranteeing reproducibility and ensuring maximum column lifetime. Available in UltiMetal for the highest operating temperatures.
- Catalog No:
- CACP7684
- CAAGCP8677
- CAAGCP7170
- CAAGCP7140
- CAAGCP8674
- CAAGCP7760
- CAAGCP7690
- CAAGCP7135
- CAAGCP7685
- CAAGCP7680
- CAAGCP7687
- CAAGCP8780
- CAAGCP7640
- CAAGCP7675
- CAAGCP7682
- CAAGCP7670
- CAAGCP8741
- CAAGCP6666
- CAAGCP7739
- CAAGCP7750
- CAAGCP6670
- CAAGCP7709
- CAAGCP7475
- CAAGCP8770
- CAAGCP7710
- CAAGCP7625
- CAAGCP8690
- CAAGCP7693
- CAAGCP8735
- CAAGCP7692
- CAAGCP8870
- CAAGCP8688I5
- CAAGCP8688
- CAAGCP7529
- Polarity:
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
Frequently Bought Together