PolarGel, Agilent Technologies
PolarGel, Agilent Technologies
PolarGel, Agilent Technologies
PolarGel GPC columns are designed for use with polar organic solvents and water/organic mixtures.
  • Proprietary surface chemistry is compatible with a wide range of polar solvents, modifiers, and mixtures
  • Excellent performance for polymers which dissolve in DMF, DMSO, NMP, DMAc, and THF/water mixtures
PolarGel GPC columns are designed for use with polar organic solvents such as DMF, DMSO, NMP, and DMAc, as well as water/organic mixtures. These eluents are very useful in GPC/SEC to separate polar polymers, such as resins, starches, acrylics, biopolymers, polysaccharides, polyamides, and other polar polymers that are insoluble in water or organic solvents.
  • Catalog No:
  • CAAGPL11171830
  • CAAGPL11171800
  • CAAGPL11176830
  • Max. pressure:
  • 150 bar
  • 150 bar
  • 150 bar
  • pH range:
  • 2-10
  • 2-10
  • 2-10
  • Temperature limit:
  • 80 °C
  • 80 °C
  • 80 °C
  • Wetted materials:
  • 0.02% Sodium azide
Product Family Options
  • Désignation
    Type de colonne
    Phase
    Granulométrie
    Ø int.
    Longueur
    Exclusion Limit
    Packing Material
  • Colonne de garde
    PL PolarGel-L
    8 µm
    7.50 mm
    50 mm
    0 - 60000 Da
    Fully Porous
    Each
    Each - $1,020.00
    $1,020.00
    Each
       $1,020.00
     : PL1117-1830
    Specifications
    Cat. No.
    CAAGPL11171830
    Max. pressure
    150 bar
    pH range
    2-10
    Temperature limit
    80 °C
    Wetted materials
  • Colonne de garde
    PL PolarGel-M
    8 µm
    7.50 mm
    50 mm
    0 - 500000 Da
    Fully Porous
    Each
    Each - $1,020.00
    $1,020.00
    Each
       $1,020.00
     : PL1117-1800
    Specifications
    Cat. No.
    CAAGPL11171800
    Max. pressure
    150 bar
    pH range
    2-10
    Temperature limit
    80 °C
    Wetted materials
  • PL PolarGel-L
    Analytique
    PL PolarGel-L
    8 µm
    7.50 mm
    300 mm
    100 - 60000 Da
    Fully Porous
    Each
    Each - $3,550.00
    $3,550.00
    Each
       $3,550.00
     : PL1117-6830
    Specifications
    Cat. No.
    CAAGPL11176830
    Max. pressure
    150 bar
    pH range
    2-10
    Temperature limit
    80 °C
    Wetted materials
    0.02% Sodium azide