InfinityLab Poroshell 120 for NP/SFC, Agilent Technologies
Fournisseur: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificats
À propos de cet article
Modern column technology based on superficially porous particles for normal phase LC separations.
- Superficially porous, 'core-shell' particles that provide both improved throughput and higher resolution
- A scalable family of particles—1.9 μm, 2.7 μm, and 4 μm—enables you to get the best from your methods and instruments
- Tested in SFC so you can benefit from faster separations and sharper peaks and worry even less about system pressures
- The InfinityLab Poroshell 120 HILIC has been highlighted as one of the most universal, best-in-class superficially porous SFC columns in independent research publications
- Excellent lot-to-lot reproducibility for confidence in your separation results
- Part of the InfinityLab family – designed for ease-of-use and maximum compatibility with Agilent instruments and supplies
- UHPLC guard columns to further extend the life of your analytical column
- Superior peak shape for faster, more accurate results due to high-purity silica and advanced bonding chemistries
- Enhanced usability, traceability, and security with a preprogrammed ID tag
Agilent InfinityLab Poroshell 120 superficially porous columns for normal phase HPLC and supercritical fluid chromatography (SFC) offer exceptional efficiency and reliability. Unique, chemistries including Poroshell 120 EC-CN, HILIC, and HILIC-Z (HILIC phases in SFC only), are available in up to three particle diameters, 1.9 μm, 2.7 μm, and 4 μm. These options provide a broad range of selectivity for your analytical method development and are scalable from traditional HPLC to UHPLC and through to ultralow dispersion UHPLC.
Spécifications
- Catalog No:
- CAAG695675-301
- CAAG693975-301
- CAAG693775-901
- CAAG695975-901
- CAAG695775901T
- CAAG695975-301
- CAAG699675-901
- CAAG699975-901
- CAAG823750-926
- CAAG695770-901
- CAAG699775-901
- CAAG693975-901
- CAAG821725-926
- CAAG693770-901
- CAAG695675-901
- CAAG821725-944
- CAAG693675-901
- Carbon load:
- 0
- 0
- 0
- 0
- 0
- 0
- 0
- 0
- 0
- 0
- 0
- 0
- 0
- 0
- 0
- 0
- 0
- Endcapped:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Max. pressure:
- 1300 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 1300 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 1300 bar
- 1300 bar
- 1300 bar
- pH range:
- 0-8
- 0-8
- 0-8
- 0-8
- 0-8
- 0-8
- 0-8
- 0-8
- 0-8
- 0-8
- 0-8
- 0-8
- 0-8
- 0-8
- 0-8
- 0-8
- 0-8
- USP code:
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- Wetted materials:
- Acetonitrile/Water