J&W HP-PONA Columns, Agilent Technologies
Specially configured for the analysis of petroleum process products, including paraffins, isoparaffins, aromatics, napthalenes, and olefins.
- 100% Dimethylpolysiloxane
- Configured for the analysis of petroleum process products
- Tested to ensure the resolution of m-xylene from p-xylene and of cyclopentane from 2,3-dimethylbutane
- PONA, PIANO
- High resolution
- Bonded, crosslinked, and solvent rinsable
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
- Similar phases: Petrocol DH, SPB-1, 007-1, Rtx-1, MXT-1, Rtx-1PONA, Rtx-DHA
Agilent J&W HP-PONA columns are configured to deliver excellent, high-resolution performance for the detailed analysis of petroleum process products, including paraffins, isoparaffins, aromatics, napthalenes, and olefins (PIANO). These 100% dimethylpolysiloxane columns are bonded, crosslinked, and solvent rinsable, and are tested to strict industry QC standards during manufacture, and to ensure the resolution of m-xylene from p-xylene and of cyclopentane from 2,3-dimethylbutane.
- Catalog No:
- CAAG19091S001E
- CA19091S-001
- Polarity:
- Low-polarity stationary phase
- Low-polarity stationary phase
Frequently Bought Together