Crosslab DB-HT Simulated Distillation Columns (moved), Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Low-bleed GC columns with a 'boiling point' phase for high-temperature simulated distillation.
- 100% Dimethylpolysiloxane
- 'Boiling point'mphase for high-temperature simulated distillation
- Durable stainless steel tubing
- 430 °C upper temperature limit
- Distillation range of C6 to C110+
- Low bleed, even at 430 °C
- Bonded, crosslinked, and solvent rinsable
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column"
Agilent J&W DB-HT Simulated Distillation columns feature a 'boiling point' phase for high-temperature simulated distillation, and durable stainless steel tubing for consistently excellent performance. These low-bleed columns have a distillation range of C6 to C110 with an operating temperature range of –60 to 430 °C. The columns are bonded, crosslinked, and solvent rinsable, and are rigorously tested to some of the strictest industry QC standards to ensure accurate results.
Specifications
- Catalog No:
- CA145-1001
- CA145-1009
- Polarity:
- Low-polarity stationary phase
- Low-polarity stationary phase