DB-1ms Columns, Agilent Technologies
À propos de cet article
An excellent general-purpose GC column, ideal for GC/MS of active analytes commonly found in drug screening and environmental analyses.
- 100% Dimethylpolysiloxane
- Identical selectivity to DB-1
- Nonpolar
- Very low bleed characteristics, ideal for GC/MS
- Improved acid performance compared to standard 100% dimethylpolysiloxane columns
- Improved signal-to-noise ratio for better sensitivity and mass spectral integrity
- Bonded, crosslinked, and solvent rinsible
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
- Similar phases: SPB-1, Rtx-1, BP-1, OV-1, OV-101, 007-1(MS), SP-2100, SE-30, ZB-1, AT-1, MDN-1, ZB-1, ZB-1ms
Agilent J&W DB-1ms is a nonpolar, low-bleed column, with an extended upper temperature limit and excellent inertness for acids and bases. The low-bleed stationary phase reduces column contribution to background noise, resulting in a higher signal-to-noise ratio and better sensitivity for improved mass-spectral purity and accurate library identification.
Bonded and crosslinked, this highly inert column provides maximum response for active analytes commonly found in drug screening and environmental analyses.
14 Options Available Below
Product Details & Documents
An excellent general-purpose GC column, ideal for GC/MS of active analytes commonly found in drug screening and environmental analyses.
- 100% Dimethylpolysiloxane
- Identical selectivity to DB-1
- Nonpolar
- Very low bleed characteristics, ideal for GC/MS
- Improved acid performance compared to standard 100% dimethylpolysiloxane columns
- Improved signal-to-noise ratio for better sensitivity and mass spectral integrity
- Bonded, crosslinked, and solvent rinsible
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
- Similar phases: SPB-1, Rtx-1, BP-1, OV-1, OV-101, 007-1(MS), SP-2100, SE-30, ZB-1, AT-1, MDN-1, ZB-1, ZB-1ms
Agilent J&W DB-1ms is a nonpolar, low-bleed column, with an extended upper temperature limit and excellent inertness for acids and bases. The low-bleed stationary phase reduces column contribution to background noise, resulting in a higher signal-to-noise ratio and better sensitivity for improved mass-spectral purity and accurate library identification.
Bonded and crosslinked, this highly inert column provides maximum response for active analytes commonly found in drug screening and environmental analyses.
Specifications for Product Family
Spécifications
- Catalog No:
- CA127-0112
- CAAG123-0131
- CAAG127-0122
- CAAG1270112LTM
- CA122-0112
- CA122-0132
- CA122-0162
- CA123-0132
- CAAG123-0112
- CAAG123-0162
- CAG3900-65002
- CA128-0112
- CA128-0122
- AG127-0123
- Polarity:
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- USP code:
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
Spécifications
Product Family Options
Informations sur le produit
- DésignationType de colonnePhaseColumn HardwareÉpaisseur du filmØ int.LongueurAvailabilityPrice
- Supplier Number: 127-0112Spécifications:
More
Specifications
Cat. No.CA127-0112PolarityLow-polarity stationary phaseUSP codeG1, G2, G9, G38 - GC column, 7" cageCapillaireDB-1msFused silica0.1 µm0.32 mm30 mLog in pour la disponibilitéEach$1,260.00Supplier Number: 123-0131Spécifications:
More
Specifications
Cat. No.CAAG123-0131PolarityLow-polarity stationary phaseUSP codeG1, G2, G9, G38 - GC column, 7" cageCapillaireDB-1msFused silica0.1 µm0.10 mm20 mLog in pour la disponibilitéEach$1,200.00Supplier Number: 127-0122Spécifications:
More
Specifications
Cat. No.CAAG127-0122PolarityLow-polarity stationary phaseUSP codeG1, G2, G9, G38 - LTM column moduleCapillaireDB-1msFused silica0.1 µm0.10 mm10 mLog in pour la disponibilitéEach$2,430.00Supplier Number: 127-0112LTMSpécifications:
More
Specifications
Cat. No.CAAG1270112LTMPolarityLow-polarity stationary phaseUSP codeG1, G2, G9, G38 - Supplier Number: 122-0112Spécifications:
More
Specifications
Cat. No.CA122-0112PolarityLow-polarity stationary phaseUSP codeG1, G2, G9, G38 - Supplier Number: 122-0132Spécifications:
More
Specifications
Cat. No.CA122-0132PolarityLow-polarity stationary phaseUSP codeG1, G2, G9, G38 - Supplier Number: 122-0162Spécifications:
More
Specifications
Cat. No.CA122-0162PolarityLow-polarity stationary phaseUSP codeG1, G2, G9, G38 - Supplier Number: 123-0132Spécifications:
More
Specifications
Cat. No.CA123-0132PolarityLow-polarity stationary phaseUSP codeG1, G2, G9, G38 - GC column, 7" cageCapillaireDB-1msFused silica0.25 µm0.32 mm15 mLog in pour la disponibilitéEach$767.00Supplier Number: 123-0112Spécifications:
More
Specifications
Cat. No.CAAG123-0112PolarityLow-polarity stationary phaseUSP codeG1, G2, G9, G38 - GC column, 7" cageCapillaireDB-1msFused silica0.25 µm0.32 mm60 mLog in pour la disponibilitéEach$2,160.00Supplier Number: 123-0162Spécifications:
More
Specifications
Cat. No.CAAG123-0162PolarityLow-polarity stationary phaseUSP codeG1, G2, G9, G38 - Supplier Number: G3900-65002Spécifications:
More
Specifications
Cat. No.CAG3900-65002PolarityLow-polarity stationary phaseUSP codeG1, G2, G9, G38 - Supplier Number: 128-0112Spécifications:
More
Specifications
Cat. No.CA128-0112PolarityLow-polarity stationary phaseUSP codeG1, G2, G9, G38 - Supplier Number: 128-0122Spécifications:
More
Specifications
Cat. No.CA128-0122PolarityLow-polarity stationary phaseUSP codeG1, G2, G9, G38 - Supplier Number: 127-0123Spécifications:
More
Specifications
Cat. No.AG127-0123PolarityLow-polarity stationary phaseUSP codeG1, G2, G9, G38