Diamond® APEX™ Pipettors, Multi-Channel, Globe Scientific
The Diamond® APEX™ 8-channel and 12-channel pipettors include the myriad of features found in the single channel models and more.
- Each channel is calibrated individually
- Removable, fully autoclavable and UV resistant manifold
- Three-year warranty
- Magnet assisted piston for extremely high accuracy and consistency
- Ergonomic design and light weight
- Precise volume adjustment
- Locking mechanism to prevent accidental volume changes
Simple operation and light weight aid in reducing fatigue and a 360° rotating manifold allows for positioning vertically or horizontally while aspirating or dispensing. As with the single channel models, every multi-channel Diamond® APEX™ pipettor is calibrated in an ISO 17025 accredited facility and come standard with a three-year warranty.
Frequently Bought Together