J&W Ultra Inert DB-624 Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Designed for trace analysis in environmental, pharmaceutical, and other volatile organic compound (VOC) applications.
- Environmental volatile organic compounds (VOCs) methods
- Excellent for US EPA Methods: 501.3, 502.2, 503.1, 524.2, 601, 602, 8010, 8015, 8020, 8240, and 8260
- Industrial chemical analysis – solvents, petrochemicals, specialty chemicals
- Food and beverage – alcohols, fusel oils
- Pharmaceutical residual solvents per USP
- Ultra inertness processing expands application range with excellent peak shape for low molecular weight acidic compounds
- UI testing ensures premium performance column-to-column
- Identical selectivity to the industry standard DB-624 – upgrade with no change in method required
- Optimized by the inventors of DB-624
Agilent J&W DB-624 UI is optimal for the fast analysis of volatile organic compounds (VOCs) in pharmaceutical, environmental, food, and industrial chemical applications. The unique Ultra Inert deactivation process enhances peak shape, improving signal-to-noise levels and increasing sensitivity for qualitative and quantitative analysis.
Ultra inertness processing expands application range with excellent peak shape for low molecular weight acidic compounds. UI testing ensures premium performance column-to-column. Identical selectivity to the industry standard DB-624 – upgrade with no change in method required.
Specifications
- Catalog No:
- CA121-1324UI
- CA125-1334UI
- CA123-1334UI
- CAAG127-1314UI
- CA122-1364UI
- CA122-1334UI
- CAAG123-1364UI
- CAAG1221364UNT
- CAAG125-1374UI
- CAAG1231364UNT
- CAAG1221334UNT
- CAAG1231334UNT
- Polarity:
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase