J&W DB-624 Columns, Agilent Technologies
Fournisseur: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificats
À propos de cet article
Recommended for EPA methods 502.2 and 8021, as well as for fast GC/MS analysis of volatiles.
- Specifically designed for the analysis of volatile priority pollutants and residual solvents
- Bonded, crosslinked, and solvent rinsable
- Equivalent to USP Phase G43
- Exact replacement of HP-624
- Available with DuraGuard (GC column with a built-in guard column)
- No cryogenics needed for U.S. EPA Method 502.2
- Excellent for U.S. EPA Methods 501.3, 502.2, 503.1, 524.2, 601, 602, 8010, 8015, 8020, 8240, 8260 and USP 467
- Excellent inertness for active compounds
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
Agilent J&W DB-624 is recommended for EPA methods 502.2 and 8021, as well as for fast GC/MS volatiles analysis. It is specifically designed for the analysis of volatile priority pollutants and residual solvents.
Spécifications
- Catalog No:
- CA122-1334
- CA122-1334E
- CAAG1251364KEY
- CAAG124-1374
- CA76514-550
- CAAG125-1314
- CAAG124-1334
- CA76514-552
- CA125-1334E
- CA128-1324
- CAAG1231364KEY
- CAAG1221364KEY
- CAAG1221334KEY
- CA125-1374
- CA125-1374E
- CA125-1364
- CAAG128-1324E
- CAAG1251334KEY
- CA123-1364
- CAAG1231334KEY
- CA123-1364E
- CA122-1364
- CA123-1334E
- CAAG125-1364E
- CA122-1364E
- CAAG125-13B4
- CAAG121-1324E
- CAAG125-13A4
- Polarity:
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- USP code:
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43